Would you like to log in again?
VWR® Critical Print™ Cleanroom Spiral Notebooks
Supplier: VWR
VWR® Critical Print™
P1114-11G P1114-12TS P1114-10C P1114-10G P1114-10-CB P1114-11C
51280-883CS 422.22 USD InStock
51280-883 51280-882 51280-916 51280-888 51280-887 51280-824
VWR® Critical Print™ Cleanroom Spiral Notebooks
Notebooks
Notebooks feature semi-transparent, high-density polyethylene covers that withstand harsh chemical vapors.
  • Low particulate level
  • Semi-transparent polyethylene cover
Low-particulate, 104g/m2 (28#) bond paper. Each notebook contains 50 sheets.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty