Would you like to log in again?
Whatman™ pH Indicator Strips, Color Bonded, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
2613-991 2614-991 2613-991 2613-991
28297-426PK 49.32 USD InStock
28297-426 51350-100 CAX28297426GSK CA28297-426
Whatman™ pH Indicator Strips, Color Bonded, Whatman products (Cytiva)
Test Paper pH Indicator Papers and Test Strips
Can be used for the rapid determination of pH values and has led to many applications in laboratories and industry
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
Individual plastic support strips carry four different segments of dye-impregnated indicator papers. The resulting combination of colour differences gives an extremely clear and accurate visual pH value. All the dyes are chemically bonded to the paper and cannot be leached into solution; problems associated with contamination of the sample and resultant anomalous readings are avoided.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty