Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Advanced® Bilirubin STAT-Analyzer™, Model BR2, Advanced Instruments

Supplier: Advanced Instruments

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Advanced® STAT-Analyzer™
BR2 B2A101 BR2 B2A102
52197-562EA 18429.5 USD InStock
CA52197-562 52197-562 52197-560 52197-566
Advanced® Bilirubin STAT-Analyzer™, Model BR2, Advanced Instruments
Bilirubinometers
Analyzer provides a rapid and accurate determination of total and direct bilirubin in neonates from a single 30µL serum sample

Using spectrophotometric analysis, total bilirubin level is determined immediately and direct bilirubin level is determined in about two minutes. Analyzer is easy to use, with simple set up, calibration, and operation.

Dimensions: 330×280×180 mm (13×11×7")

Certifications: ETL-C listed. CE marked. Analyzer test system is categorized as a CLIA non-waived test.

Ordering information: Manufacturer’s one-year warranty on workmanship and all parts except glass, plastic, and parts under warranty by their manufacturer.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty