Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Cryoscope Standards, Advanced Instruments
Supplier: Advanced Instruments
3LA022 3LA033 3LA023
52198-144EA 63.92 USD InStock
52198-144 55005-001 55005-003
Cryoscope Standards, Advanced Instruments
Standards Environmental Analysis Standards
Cryoscope standards are used to calibrate and/or check the performance of a cryoscope.
Standards should only be used in accordance with the cryoscope being calibrated. Refer to the specific instrument's user guide for calibration instructions. These standards are prepared from salts. 2-Hydroxybiphenyl Sodium Salt Tetrahydrate, 0.01%, is added as a preservative.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty