Would you like to log in again?
VWR® Signature™ High-Performance Horizontal Air Flow Ovens
Supplier: Avantor
VWR® Signature™
1690 1680 1685
52202-509EA 22112.77 USD InStock
52202-509 52202-203 52202-258
VWR® Signature™ High-Performance Horizontal Air Flow Ovens
Ovens Standard Ovens
Horizontal airflow is created using a turbo blower that re-circulates air over the heating element, providing fast heat-up and recovery times, and ensuring a more rapid and efficient rate of drying.
  • True horizontal airflow design
  • Programmable 24-step ramp and soak controller
  • Type-304 stainless steel interior and exterior
  • Durable powder-coat paint exterior finish
  • Three-inch intake and exhaust ports
  • Equipped with adjustable stainless steel shelves
  • VWR® Two-Year Limited Parts and Labor Warranty
By forcing continuously warmed air into the chamber, the turbo blower assures uniform heating across all shelves and to every area of the chamber, eliminating concerns of obstructed air, regardless of how the shelves are loaded.
To maintain temperature uniformity and stop heat loss, a 8.9 cm (3¹/₂") thick, double-layer insulating barrier of high-density alumina silicate and high-temperature fiberglass wraps around all five sides of the chamber. A high temperature silicone rubber door seal eliminates air leakage. In addition, the door latch and hinges are user-adjustable to maintain a better seal as metal expands and contracts over the life of the oven.
The independent over-temperature safety is user-adjustable and protects the unit against damage to contents and burnout in the unlikely event that the main controller fails. The sealed membrane touch-pad control panel guards against any harm to the electronics.
Stainless steel interior and exterior provide maximum corrosion resistance. Exterior dimensions include handle and exhaust port.
Ordering information: Supplied with adjustable stainless steel shelves.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
COMPLEMENTARY PRODUCTS
SERVICES
Services: VWR CATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
White Glove Installation and Verification Service–the white glove installation includes unpacking, inspection, removal/disposal of packing materials, transporting order via elevator and merchandise set up in the room of your choice, installation of casters or leveling feet, power and electrical connections inspected, supply voltage verified, functional and performance verification documented. Our engineers will move the unit from the customer's receiving dock to the lab within the same building. This service does NOT include transportation from vendor warehouse to customer's receiving dock.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty