Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crossed Blade Impeller, 2", Caframo

Supplier: Caframo
A130 A130
52428-768EA 147.34 USD InStock
CA58960-140 52428-768
Crossed Blade Impeller, 2", Caframo
Stirrers Stirring Tools
The Caframo Crossed Blade Impeller features a 2" diameter, and is welded to a 5/16" diameter by 16" shaft

The impeller will vortex 5 gallons of water at 900rpm. Radial flow.

Composed of 316L electropolished stainless steel, it offers very high shear with high vortexing characteristics.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty