Would you like to log in again?
Anaerobic PEA, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A90
52428-860PK 23.7 USD InStock
52428-860
Anaerobic PEA, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Anaerobic PEA (CDC formula), reducible, for anaerobes, 15x100mm plate, order by the package of 10, by Hardy Diagnostics
(CDC formula with phenylethanol, 5% sheep blood, hemin, and vitamin K) For the isolation of anaerobic bacteria.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty