Would you like to log in again?
Accessories for Oxygen Meters, YSI
Supplier: YSI
052390 098095 057087 098088 59854 052391 98094 098400 052392
52457-208EA 516.44 USD InStock
52457-208 52457-610 52457-585 52457-552 52457-212 52457-066 52457-210 52457-064 52457-010
Accessories for Oxygen Meters, YSI
Electrochemical Meters Oxygen Meters
Oxygen-temperature probe 52457-010 features a pressure compensating design that ensures error-free deep-water operation
Supplied with membranes, KCI solution, and waterproof connector for cables. Submersible stirrer agitates sample directly in front of probe membrane. Stirrer is powered by the meter battery pack, and includes a 15.2m (50') cable. Cable assemblies connect the probe to the meter. B.O.D. non-stirring bottle probe includes an oxygen electrode only, and features a probe designed to fit the neck of a B.O.D. bottle. Field dissolved oxygen (D.O.) probes include six cap membranes and electrolyte solution. Membrane kits contain KCI solution and membranes. The O-ring kit includes 24 O-rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty