Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dissolved Oxygen Cap Membrane Kit, YSI

Supplier: YSI
059880
52457-598EA 140.45 USD InStock
52457-598
Dissolved Oxygen Cap Membrane Kit, YSI
Filters Capsule Filters
Membranes are 1 ml FEP fluoropolymer film. Response time (T95 - from 0 to 100%) is 18 seconds.

  • Caps are black
  • Required flow rate is 12" per second

Flow dependence after 4 minutes is 45%.

Delivery information: Kit includes one bottle of electrolyte, one sanding disc, and six screw-on cap membranes.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty