Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters, Thermo Scientific
Supplier: Thermo Scientific
Orion™
80113 80513
52458-106EA 233.24 USD InStock
52458-106 34104-430
Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters, Thermo Scientific
Electrodes Dissolved Oxygen Electrodes
These probe maintenance kits help ensure optimal performance of Orion™ polarographic dissolved oxygen probes.
Ordering information: Each probe maintenance kit includes two membrane caps, one electrolyte solution, and a polishing disk.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty