Would you like to log in again?
Whatman™ SPARTAN Syringe Filters Certified for HPLC Sample Prep, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10463042 10463053 10463100 10463032 10463040 10463112 10463060 10463112 10463050 10463040 10463052 10463102 10463053 10463062 10463030 10463100 10463050 10463102 10463032 10463052 10463062 10463110 10463030 10463110 10463060 10463042
97005-220PK 1397.45 USD InStock
CA97005-230 97005-220 CA97005-232 97005-222 CA97005-234 CA52844-780 97005-224 CA97005-224 52844-784 CA97005-226 52844-786 CA97005-228 CA52844-776 97005-230 97005-232 97005-234 CA97005-220 52844-780 CA97005-222 52844-782 CA52844-782 97005-226 CA52844-784 97005-228 52844-776 CA52844-786
Whatman™ SPARTAN Syringe Filters Certified for HPLC Sample Prep, Whatman products (Cytiva)
Filters Syringe Filters
SPARTAN syringe filters with regenerated cellulose (RC) membrane provide reproducible filtration of organic and aqueous solutions for high performance and ultra-high performance liquid chromatography (HPLC and UHPLC) sample preparation.
  • Hydrophilic membrane made of regenerated cellulose fiber exhibits low protein binding membrane
  • Disposable filter unit with PP housing
  • Excellent chemical resistance to common aqueous and organic solvents, so recommended for HPLC method sample preparation
  • Tested and certified for the absence of UV-absorbing substances at wavelengths of 210 and 254 nm with water, methanol, and acetonitrile
  • Documented batch-to-batch quality and consistency, ensuring reproducible results
  • Low dead volume Mini-tip option
  • Recyclable after use
HPLC sample preparation often requires a filtration step before samples can be used for liquid column chromatography. This step helps to clarify peaks of chemical compounds being analyzed and protects the HPLC column from large particles that might damage it.
SPARTAN regenerated cellulose filters from Cytiva's business cater to this need, because their regenerated cellulose hydrophilic membrane is resistant to common HPLC grade solvents and exhibits very low levels of extractables that could otherwise interfere with chromatographic analysis.
In addition to their use as HPLC syringe filters, SPARTAN filters can be used in capillary gel electrophoresis, protein sample ultrafiltration, UV/VIS analysis, and any application requiring a chemically resistant, low protein binding, hydrophilic membrane in a polycarbonate filter housing.
Certifications: Certified for HPLC sample prep.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty