Would you like to log in again?
PYREX® Moisture Test Distilling Receiver, Corning
Supplier: Corning
PYREX®
3611-25 3611-10
52929-367CS 396.14 USD InStock
52929-367 52929-334
PYREX® Moisture Test Distilling Receiver, Corning
Distillation Components Distillation Receivers
Receivers, made of PYREX® brosilicate glass, are designed for determining water by distillation.
  • Have 24/40 standard taper ground joint on the sidearm
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
The 10 ml receiver is graduated from 0 to 1 ml in 0.1 ml increments, with a maximum scale error of 0.05 ml. The graduation intervals from 1.0 to 10.0 ml are 0.2 ml, with maximum scale error of 0.1 ml. The 25 ml receiver is graduated from 0 ml to 1 ml, in 0.1 ml increments, with a maximum scale error of 0.05 ml. The graduation intervals from 1.0 to 25.0 ml are 0.2 ml, with a maximum scale error of 0.1 ml.
Certifications: ASTM E-123.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty