Would you like to log in again?
Transfertube® Disposable Harvesters, Spectrum® Laboratories
Supplier: Spectrum
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Transfertube®
190195 190195P
52950-160CS 1371.27 USD InStock
52950-160 52950-159
Transfertube® Disposable Harvesters, Spectrum® Laboratories
Samplers Fluid Samplers
Designed for quick and easy, one-hand transfer of agar plugs and the harvesting of microbial colonies
Just insert harvester, pull plunger, transfer, and depress plunger to release. Also recommended for taking liquid samples. Comes as a complete unit that does not require a rubber bulb. Holds approximately 4mL of liquid. Autoclavable. Made from polypropylene.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty