Would you like to log in again?
PYREX® Volumetric Pipettes, Color-Coded, Class A, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PYREX®
7100-5 7100-10 7100-2 7100-20 7100-3 7100-25 7100-100 7100-4 7100-50 7100-1 7100-15
53044-172CS 383.66 USD InStock
53044-172 53044-070 53044-194 53044-092 53044-140 53044-059 53044-037 89091-078 53044-117 53044-139 89091-456
PYREX® Volumetric Pipettes, Color-Coded, Class A, Corning
Pipettes Graduated Pipettes
Colored markings remain sharply legible for the life of pipet.
  • Reusable, rugged construction
  • Color-coded
  • Colored enamel graduations and markings
Ruggedly constructed and properly proportioned, pipettes have accuracy tolerances to meet or exceed the strict requirements for Class A Volumetric Ware as indicated by ASTM E969.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty