Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
PYREX® Measuring (Mohr) Pipettes, Color-Coded, Tempered Top and Tip, Colored Markings, Corning
Supplier: Corning
PYREX®
7065-1X 7065-1 7065-2 7065-10 7065-5 7065-25 7065-1C
53083-006CS 233.99 USD InStock
53083-006 53083-039 53083-120 53083-119 53083-072 53083-094 53083-050
PYREX® Measuring (Mohr) Pipettes, Color-Coded, Tempered Top and Tip, Colored Markings, Corning
Pipettes Measuring (Mohr) Pipettes
These reusable glass pipettes are color-coded by size for easy identification and sorting.
  • Top end is constricted (Mohr type)
  • Colored graduations are enamelled onto the glass
  • Calibrated 'to deliver' (TD) their total capacity without blow-out
Sharp, fine lines provide accurate reading. Large numbers are easy to read. The top end of 5 to 25 ml sizes is constricted.
Certifications: ASTM E-1293.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty