Would you like to log in again?
Pure™ 10 Pipette Tips, Molecular BioProducts
Supplier: Molecular BioProducts
Pure™
3500 3502
53225-660CS 1083.22 USD InStock
53225-660 53225-662
Pure™ 10 Pipette Tips, Molecular BioProducts
Pipette Tips
These pipette tips are ideal for low volume pipetting.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Nonsterile tips are autoclavable
The MicroPoint* design increases precision when working with low volume samples as the tip profile is nearly 50% smaller than standard blunt-end tips and much more narrow than beveled-end tips. Tips are ideal for a wide variety of applications and are compatible with Pipetman*, Nichiryo*, Biohit*, Finnpipette*, Rainin*, and Transferpette* pipettors.
Ordering information: Tips 53225-664 and -662 are packaged 96 per tray, 10 trays per pack. Tips 53225-660 are bulk packaged.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty