Would you like to log in again?
VWR® Culture Tubes, Disposable, with Screw-Cap Finish, Borosilicate Glass
Supplier: VWR
VWR®
23283.800 23283.806 23283.685 23283.686 23283.804 23283.802
53283-806CS 892.66 USD InStock
53283-806 53283-685 53283-686 53283-800 53283-802 53283-804
VWR® Culture Tubes, Disposable, with Screw-Cap Finish, Borosilicate Glass
Tubes Culture Tubes
Tubes have a marking spot and reference mark for orientation during use and a threaded neck to accept screw cap.
  • Borosilicate glass constuction
  • Marking spot and orientation reference mark on all tubes
Delivery information: Caps are not supplied, sold seperately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty