Would you like to log in again?
PYREX® Serological Pipettes, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PYREX®
7077B-25 7078B-25 7078-1N 7078B-50 7077B-50 7077-2N 7078D-1 7079-5N 7078-2N 7077-5N 7078D-10 7077B-1 7079-1N 7078B-1 7079-2N 7077B-10 7079-10N 7078-1CN 7078B-5 7078-5N 7078D-5 7078B-10 7078-10N 7077-1N 7077B-5 7077-10N
89091-106CS 615.78 USD InStock
89091-106 53284-449 53284-427 53284-347 53284-369 53284-325 53284-620 53284-463 53284-562 53284-584 53284-100 53284-121 53284-143 53284-165 53284-187 53284-460 53284-380 89091-220 89091-484 89091-120 89091-268 53224-610 89091-224 89091-488 53284-391 53284-609
PYREX® Serological Pipettes, Corning
Pipettes Serological Pipettes
These sterile disposable 1 ml PYREX® serological pipettes, calibrated (TD) with blow-out, offer long, slender, tapered tips to make pipetting go faster and easier.
  • All pipettes have negative graduations
  • Fire-polished tips for burr-free, even flow rates
Easy-open Steri-View™ wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size.
Certifications: Colors conform to I.S.O. standards.
Packaging: Non sterile pipettes available in bulk package; sterile pipettes available in individual paper-plastic wrappers or as Multi-Pack in plastic bags.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty