Would you like to log in again?
VWR® Pipette Fillers
Supplier: VWR
VWR®
5010BU 5010BL 5010GR 5010RE
53497-111CS 8559.07 USD InStock
53497-111 53497-009 53497-086 53497-053
VWR® Pipette Fillers
Pipettes Pipette Bulbs
Pipette fillers do away with the frequently hazardous practice of drawing liquids into pipettes by mouth.
  • 100% natural rubber construction
  • For use with 1 to 45 ml pipettes
  • Four color options available
They work safely with toxic, infectious, odoriferous, radioactive, corrosive, or sterile liquids.
Pipettes are made out of 100% natural rubber with three stainless steel ball valves. For use with pipettes of 1 to 45 ml capacity and 4 to 8 mm outside diameter. A volume of 100 ml is attainable with repeated evacuation. With a simple three-step process, one can eliminate dangerous mouth pipetting.
Also useful for skimming off liquids floating above a chemical precipitate or sediment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty