Would you like to log in again?
Monoject™ Blunt Cannulas, Covidien
Supplier: Covidien
Monoject™
8881202389 8881202397 8881202348 8881202371 8881202314 8881202355 8881202330 8881202322 8881202363
15141-142BX 88.28 USD InStock
15141-142 53498-484 15141-144 53498-478 MJ8881-202355 15141-146 53498-480 15141-148 53498-482
Monoject™ Blunt Cannulas, Covidien
Syringe Needles
These sterile, stainless steel cannulas feature an aluminum Luer-lock hub and a color-coded needle sheath with a charcoal gray cap
Autoclavable in rigid pack for use in O.R./procedure packs. Latex-free.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty