Would you like to log in again?
Monoject™ Syringes, Sterile, Covidien
Supplier: Covidien
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Monoject™
8881516135 8881513934 Z8881513918 8881516911 8881516937 8881535770 8881513538 8881560141 8881535762 8881520657 8881513033 8881513512 8881560224 Z8881513918 8881560125 8881520673 8881513058 8881513231 8881513256 8881520657 8881535796 8881513918
53498-496BX 60.02 USD InStock
53498-496 15141-233 15141-222 15141-091 MJ8881-535796 53498-490 53498-492 CA11010-422 53498-494 MJ8881-535762 MJ8881-513033 MJ8881-513231 MJ8881-520657 MJ8881-560141 MJ8881-513256 MJ8881-516911 15141-227 CA15141-225 MJ8881-513538 15141-224 CA15141-224 15141-236
Monoject™ Syringes, Sterile, Covidien
Syringes Syringes, Medical
These latex-free Covidien Monoject™ syringes feature a color-coded translucent needle hub and a rigid pack cap.
The 3mL, 6mL, 12mL, and 20mL syringes feature bold graduations; the 35mL and 60mL syringes feature bold dual graduations. The 35mL displays 1oz. graduations in 0.25oz. increments, and the 60mL displays 2oz. graduations in 0.25oz. increments. Syringes are autoclavable in the rigid pack for use in O.R./procedure packs.
Available with or without sterile, ultra-sharp, tri-beveled, anti-coring needles. Syringes with needles feature a Luer-lock tip for complete protection from contamination of sterile fluid pathway. Syringes without needles are available with regular, Luer-lock, catheter, eccentric Luer, or Toomey type tips. Luer-lock syringes feature a threaded tip to lock the needle in place.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty