Would you like to log in again?
Microtest Manifolds, DWK Life Sciences
Supplier: DWK Life Sciences
851388 851381
53500-336EA 782.33 USD InStock
53500-336 53500-343
Microtest Manifolds, DWK Life Sciences
Manifolds
These autoclavable stainless steel microtest manifolds are ideal for use with 24 or 96 well microplates
It tightly fit with both Luer and Luer Lock syringe nozzles. The manifolda available in both a straight "T" style and an angular style with an offset hub.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty