Would you like to log in again?
Upchurch Scientific® PEEK™ Frits, IDEX Health & Science
Supplier: IDEX Health & Science
A-700 A-701 A-735X A-722 OC-802 A-702 A-707 A-706 A-735 A-710 A-720 A-704 A-703
21511-618EA 16.5 USD InStock
21511-618 53501-226 53501-218 21511-458 21001-314 35321-094 21511-456 53501-360 53501-220 53501-362 53501-222 53501-332 53501-334
Upchurch Scientific® PEEK™ Frits, IDEX Health & Science
Filters Precolumn Filters and Frits
Upchurch Scientific® PEEK™ frits are suitable for low- and high-pressure applications and offer exceptional uniform porosity to ensure longer filtration life and consistent frit-to-frit swept volumes.
While all Upchurch Scientific® PEEK™ frits are capable of withstanding pressures >5,000 psi (>345 bar), the actual pressure holding capacity of each frit is usually dependent on the filter body in which it is placed.
Upchurch Scientific® Frit-In-A-Ferrule™ seal and filter simultaneously and can filter at any point in your system where ¹/₆ or ¹/₈" Ext.Ø tubing is used in a flat-bottom 1/4-28, M6 or 5/16-24 connection. For more information and a listing of these products, search on 53501-450.
PEEK™ polymer discs are biocompatible, metal free, and inert to most solvents. Disc rings, included on most PEEK™ frits, are made of PCTFE and also also offer excellent chemical resistance. PCTFE surrounded PEEK™ frits can be used up to 80 °C, and PEEK™ frits alone are a good choice for applications up to 100 °C.
Ordering information: The thickness dimension shown in the frit drawings represents the thickness of the frit disc, not the frit ring. Frit rings are slightly thicker to ensure a proper seal. When tightenened, into a filter holder the ring compresses to the thickness of the frit disc.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty