Would you like to log in again?
Upchurch Scientific® Inline Filters, IDEX Health & Science
Supplier: IDEX Health & Science
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A-411 M-520 VHP-505 A-424 A-430 A-410 A-429X M-120X A-416 A-330 A-428X A-431 A-314
21001-162EA 31.7 USD InStock
21001-162 53501-412 53498-596 53501-416 53498-598 53500-660 53500-662 53500-606 21511-442 53501-422 21511-450 89218-736 21511-000
Upchurch Scientific® Inline Filters, IDEX Health & Science
Filters In-Line Filters
Placed between the pump and sample injection valve, an inline solvent filter traps particles released through normal pump piston seal wear
Upchurch Scientific® inline solvent filters come in three 'families' — MicroFilter®, analytical scale, and semi-prep scale. The Upchurch Scientific® inline MicroFilter® offers extremely low internal volume and 0.5 µm filtering porosity. The MicroFilter® can be connected with a variety of capillary tubing sizes (70 to 520 µm OD) using the provided fittings and Upchurch Scientific® MicroTight® tubing sleeves. Please search on product number 53403-002 for a selection of MicroTight® tubing sleeves.
Analytical scale inline solvent filter products can be further broken down into three categories: standard, biocompatible and those for very high pressure (VHP) applications, such as UHPLC. The standard and biocompatible lines offer easy connections to flow path tubing and a choice of frit porosities to customize the level of filtration. The special VHP inline filters can be used at pressures up to 15,000 psi (1,035 bar) and also offer a choice of frit porosity.
For an alternative to traditional inline filters, try Upchurch Scientific® Frit-In-A-Ferrules™. For more information and a listing of these products, search on product number 53501-430.
Semi-Prep Inline Filters feature larger frit surface areas, larger thru-holes, and larger overall volumes to accommodate the higher flow rates involved in analyses of this scale. Both standard and biocompatible options are available.
If left suspended in the fluid pathway, these particles would be flushed through to the sample injection valve, resulting in valve damage and further system contamination
Certifications: ROHS Certified, excluding 89218-734, 89218-738, and 89218-740.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty