Would you like to log in again?
Upchurch Scientific® Specialty Barbed Adapters, IDEX Health & Science
Supplier: IDEX Health & Science
Upchurch Scientific®
P-767 P-799 P-757
21511-622EA 93.91 USD InStock
21511-622 53501-470 21511-230
Upchurch Scientific® Specialty Barbed Adapters, IDEX Health & Science
Fittings and Connectors
These unique Upchurch Scientific® tubing adapters connect peristaltic tubing to standard 1/16" or 1/8" OD tubing (using different fittings than are supplied with the product, available separately).
  • Easy to use
  • Specially designed nose is manufactured from inert PEEK™ polymer
  • Can be used as "stops" for peristaltic pump
Manufactured from inert PEEK™ polymer, a specially-designed nose allows the peristaltic tubing to simply press fit over the nose and then be held tightly in place by the retainer sleeve
These Upchurch Scientific® Conical Adapters, designed for peristaltic tubing, provide a reliable connection between rigid/semi-rigid tubing and peristaltic-type flexible tubing, such as Tygon® and PharMed®. Composed of a PEEK™ polymer female nut, a Super Flangeless™ ferrule system and an ETFE or PEEK conical adapter body, the narrow coned end of the adapter body allows peristaltic tubing to slide on more easily than it does onto conventional barbed adapters. Peristaltic tubing is also easier to remove from Conical Adapters since no cutting or excessive pulling is required.
1/16" OD tubing may then be connected with the Flangeless Fittings supplied with the adapter. To connect peristaltic tubing to 1/8" OD tubing, simply replace the supplied fittings with your choice of Flangeless Fittings. One popular application for these adapters is to use them as "stops" for your peristaltic pump. By doing so, you can reduce the amount of peristaltic tubing required for your flow path, thus reducing cost.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty