Would you like to log in again?
SOCOREX® LAB 45™ Reagent Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
SOCOREX® LAB 45™
844030 W832010 844027 844026 844025 W832006 W832014 W832012 W832008 844029 W832016 W832000 W832004
89044-830EA 182.07 USD InStock
89044-830 53502-519 89044-824 53502-517 89044-826 89044-816 89044-820 89044-822 53502-520 53502-552 53502-550 89044-828 89044-818
SOCOREX® LAB 45™ Reagent Bottles, DWK Life Sciences
Bottles Glass Bottles
Selection of glass reagent bottles, each supplied with one screw cap.
  • Clear bottles are made from borosilicate glass
  • Amber bottles are square, round or with jug handle
Perfectly adapted with Calibrex™ bottle top dispensers. Corresponding neck adapters supplied with the dispensers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty