Would you like to log in again?
Dialamatic Microdispensers, Drummond
Supplier: Drummond
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3-000-210 3-000-275 3-000-225 3-000-250
53507-378PK 538.75 USD InStock
53507-378 53506-278 53506-274 53506-270
Dialamatic Microdispensers, Drummond
Pipettes Repetitive Pipettors
Dialamatic Microdispensers feature borosilicate glass bores with stainless steel plungers.
  • Borosilicate glass bores with stainless steel plungers (PTFE tips on all plungers 25 µl or larger)
  • Adjustable, direct reading plunger sets volume
  • Delivery scale engraved on plunger shaft
  • Fractional deliveries possible with pre-marked indexes
  • Volumetric accuracy of ±1% at full volume
The microdispensers have a volumetric accuracy of ±1% at full volume.
The direct reading plunger sets the volume, and the plunger shaft includes delivery-scale engravings.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty