Would you like to log in again?
Fixed Volume Microdispensers, Drummond Scientific
Supplier: Drummond
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3-000-320 3-000-375 3-000-310 3-000-325 3-000-350 3-000-390 3-000-388 3-000-105 3-000-385
53507-412PK 485.01 USD InStock
53507-412 53507-368 53507-410 53507-386 53507-384 53507-408 53507-416 53507-390 53507-406
Fixed Volume Microdispensers, Drummond Scientific
Pipettes Repetitive Pipettors
Fixed Volume Microdispensers have an accuracy of ±1% at full volume.
  • Borosilicate Glass Bores
  • Stainless Steel Plungers
  • Accuracy at Full Volume: ±1%
The microdispensers feature borosilicate glass bores with stainless steel plungers.
All plungers 25 µL or larger feature PTFE tips.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty