Would you like to log in again?
MBP DNA AWAY™
Supplier: Molecular BioProducts
7010
53509-506CS 749.39 USD InStock
53509-506
MBP DNA AWAY™
Cleaning Agents Decontaminants
RNase AWAY® eliminates RNase and DNA from laboratory surfaces
  • Easily Remove RNase Contaminants
  • Safe to Use on Pipettors, Glassware, Plasticware, and Most Metal Surfaces
  • Space-Saving, Convenient Packaging
  • Eliminates Over-Spraying of Decontaminants on Sensitive Equipment
The surface decontaminant is ideal for decontaminating apparatus, benchtops, glassware, and plasticware. The decontaminant leaves no residue to interfere with gel polymerization or staining and contains no strong acids. DNA AWAY™ decontaminant is ideal for removing DNase and DNA contamination.
Certifications: Biotechnology grade. Application Conformity Pass. Store frozen. Irritant. Hazard Statements: H315 / H319 / H335. Precautionary Statements: P280 / P302+P352 / P304+P340 / P305+P351+P338.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty