Would you like to log in again?
ART® 300 Barrier Pipette Tips, Sterile, Thermo Scientific
Supplier: Thermo Scientific ART
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ART®
2070 2070-HR 2070-05-HR
53509-508CS 1136.98 USD InStock
53509-508 89413-264 89413-268
ART® 300 Barrier Pipette Tips, Sterile, Thermo Scientific
Pipette Tips
Tips ensure 100% protection against aerosol and liquid contamination.
  • Pre-sterilized tips
  • Secure fit and extra space for repeat pipetting
The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required. Tips are compatible with Eppendorf®, Transferpette®, Finnpipette®, Biohit®, and Rainin® series pipettors.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Ordering information: Available in both traditional lift-off and versatile hinged racks.
Packaging: Sterile tips are packaged 96/tray, 10 trays per pack.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty