Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safe-Lock Tubes, 1.5 ml - Microtubes

Supplier: Eppendorf
022363204 022363301 022363204 022363212 022363280 0030123611 022363328 022363212 022363263 022363247 022363301 022363221 022363328 022363263 022600028 022363280 022600028 022363247 022363221
21008-960PK 102.76 USD InStock
CA20901-547 21008-960 76019-014 20901-547 21008-955 CA21008-953 CA21008-955 21008-957 CA21008-960 21008-961 CA53511-997 21008-953 CA21008-961 53511-997 21008-959 62111-752 CA21008-957 CA21008-959 CA62111-752
Safe-Lock Tubes, 1.5 ml - Microtubes
Tubes Microcentrifuge Tubes
Eppendorf® Safe-Lock 1.5 ml microtubes ensure secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1.5 ml capacity.

  • Hinged Safe-Lock lid prevents inadvertent lid opening during incubation and storage
  • g-Safe® technology, depending on the Safe-Lock Tube variant (refer to the manual), ensures outstanding centrifugation stability up to 30000×g
  • g-Safe® technology prevents sample loss due to tube breakage, and provides additional safety when handling hazardous samples
  • Polypropylene construction offers robust resistance to chemicals, mechanical stress, and extreme temperatures
  • Precision lid sealing minimizes evaporation rates during extended storage periods
  • Generously sized frosted lid and side surface facilitate easy labeling
  • Assured functionality in a temperature range from −86 to +100 °C
  • Autoclavable when open (121 °C, 20 min.) for reliable sterilization
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur® quality for tested purity

Eppendorf® Safe-Lock 1.5 ml microtubes exemplify secure sample storage with their patented Safe-Lock design, preventing accidental openings and preserving sample integrity. Versatile across laboratory applications, the durable construction and 1.5 ml capacity cater to varied sample volumes. Clear graduations and a smooth surface enhance user-friendliness, ensuring a seamless and reliable laboratory experience. Elevate scientific endeavors with the trusted quality of Eppendorf® Safe-Lock tubes.

Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA grade can be downloaded from the Eppendorf website.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty