Would you like to log in again?
Rotors for Vacufuge Plus
Supplier: Eppendorf
022822128 022822179 022822144 022822233 022822080 022822136 022822187 022822241 022822179 022822136 022822161 022822228 022822225 022822225 022822144 022822004 022822080 022822047 022822004 022822209 022822233 022822241 022822047 022822128
47745-388EA 1298.92 USD InStock
CA47745-386 47745-388 47745-386 53512-628 CA47745-384 CA53512-972 58922-840 53512-974 CA53512-630 CA53512-974 89213-150 CA47745-388 CA58922-840 58922-842 47746-348 CA47746-348 CA53512-628 47745-384 58922-844 53512-640 CA53512-640 53512-630 58922-846 53512-972
Rotors for Vacufuge Plus
Centrifuges Rotors
Eppendorf Vacufuge Plus efficiency meets precision. Tailor your concentration processes with a max speed of 1700 rpm. Accommodate diverse samples with a 48×1.5/2.0 ml capacity. Customize your setup with various accessories for optimal sample concentration.
  • A wide range of fixed-angle rotors and the microplate rotor are available to accommodate a variety of tubes, plates, and applications
  • Increase capacity with a spacer to stack some fixed-angle rotors
  • Precision engineered for optimal sample concentration
Revolutionize your sample concentration with the Eppendorf Vacufuge Plus. Operating at a max speed of 1700 rpm, this centrifuge concentrator offers versatility for DNA/RNA, protein purification, and more. The 48×1.5/2.0 ml capacity caters to various sample volumes, ensuring efficient concentration. Customize your workflow with accessories like rotors and adaptors. The user-friendly interface and compact design make the Vacufuge plus an essential tool for any lab. Elevate your concentration processes with precision and adaptability.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty