Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benchtop Centrifuge, 5804

Supplier: Eppendorf
022622552 022629948 022628214 022628203 022628048 022628149 022628048 022628034 022622501 022628135 022622501 022628149 022628034 022628211 5804000579 022628203 022628135 022622552
490013-046EA 13625.7 USD InStock
490013-046 CA11017-350 CA11017-354 75856-528 89213-140 CA89210-968 97007-368 CA89210-972 89210-968 53513-802 CA53513-800 53513-800 89305-166 CA53513-802 89210-972 CA89210-970 89210-970 89305-158
Benchtop Centrifuge, 5804
Centrifuges Benchtop Centrifuges
The high-speed Eppendorf Centrifuge 5804 is a medium capacity centrifuge, ideal for molecular applications in tubes up to 250 ml. It also offers supreme versatility with the capability of taking swing-bucket and fixed-angle rotors, as well as deepwell plates.

  • Low access height of 29 cm for easy loading and unloading of samples
  • Quiet operation to improve work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 to 250 ml
  • Plate rotor for centrifugation of all types of MTP, PCR, cell culture, or Deepwell Plates
  • Fixed-angle rotors for high-speed applications in tubes from 0.2 to 85 ml

Centrifuge 5804, a benchtop centrifuge renowned for its quality and reliability, offers the most cost-effective solution for medium-throughput applications requiring speeds of up to 20,913×g (14,000 rpm). Experience true versatility with a broad selection of rotors and adapters that accommodate tubes and bottles from 0.2 to 250 ml, as well as MTP, PCR and deep-well plates.

The swing-bucket rotor and adapters accommodate tubes and bottles from 0.2 to 250 ml whereas the fixed-angle rotors accomodate vessels from 0.2 to 85 ml. The plate rotor is ready for you to carry out centrifugation of all common types of multiwell plate, such as PCR, cell culture or deep-well plates.

Max. Capacity Plate: 10×MTP/2×DWP
Rotors Available: 12

Certifications: UL Listed, Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001).

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty