Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MLA Precision Pipettors, VistaLab
Supplier: VistaLab
1030 1118 1103 1111 1028 1110 1023 1027 1102 1021 1026 1109 1115 1025 1029 1113 1107 1104 1119 1022 1505 1108 1106 1024 1117 1101 1112
49000-514EA 233.33 USD InStock
49000-514 53514-014 53514-016 49000-532 53514-018 49000-530 49000-518 75813-802 53514-062 75813-804 53514-020 53514-064 53514-022 53514-066 53514-068 53514-004 49000-526 49000-524 53514-006 53514-008 49000-522 53514-070 53514-072 53514-074 75814-378 53514-010 53514-012
MLA Precision Pipettors, VistaLab
Pipettes Repetitive Pipettors
MLA pipettors are made of precision-engineered aluminum alloy and stainless steel, and are designed for highly accurate delivery and proven performance.
  • Available in a wide range of fixed volumes
  • Light plunger forces for easy operation with thumb/forefinger
  • Single-stroke 'to deliver' operation
  • Individually serialized
  • Lifetime warranty on parts
These pipettors are available in 28 fixed volumes ranging from 5 to 1000 μL. Pipettors can be easily adjusted as needed to achieve accurate performance on dense or viscous liquids.
Accessories information: Seal Kits for MLA Pipettors are available separately. Seal kits contain lubricant, seals, and instructions.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty