Would you like to log in again?
Finntip® Filtered Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
94052310 94052550 94052410 94052020 94052200 94052300 94052350 94052100 94052060 94052150 94052800 94052320 94052000 94052430 94052160 94052600
14233-402CS 300.81 USD InStock
14233-402 53515-908 14233-404 14222-044 11310-904 46600-006 76637-686 53515-910 53515-900 14224-026 53515-898 53515-984 53515-262 11388-542 46620-286 12777-872
Finntip® Filtered Pipette Tips, Thermo Scientific
Pipette Tips
Finntip Filter Pipette Tips provide superior protection against contamination.
  • Sterilized with irradiation
  • Certified free from DNA, DNase, RNase and endotoxin
Finntip filter pipette tips are available in racks with an assortment of volumes from 0.2 to 10 μl. Made from an inert non-sealing polypropylene matrix, the filter is effective at eliminating contamination from the pipetter to the sample, thereby ensuring the integrity of any pipetting function.
For 14233-402, CS of 10, there are 10 racks in a case, and each rack has 96 tips.
These sterile tips are produced in a controlled environment and ideal for a wide range of applications, especially sensitive procedures including PCR based reactions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty