Would you like to log in again?
Finntip Stepper Tips, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9404173 9404190 9404170 9404223 9404183 9404220 9404213 9404230 9404193 9404200 9404203 9404233 9404180 9404210
53515-918PK 34.16 USD InStock
53515-918 14222-046 53515-916 53515-938 53515-928 53515-936 53515-926 53515-934 53515-924 53515-932 53515-930 53515-226 53515-920 53515-940
Finntip Stepper Tips, Thermo Scientific
Pipette Tips
Finntip stepper tips are the perfect fit with Thermo Scientific Finnpipette stepper pipetters.
The finntip stepper tips are plunger-style tips for positive displacement pipetting. Seven sizes are available in sterile and non sterile versions.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty