Would you like to log in again?
BIOCONTROL TIP CONE 12CH 300UL
Supplier: Thermo Fisher Scientific
12-Channel Tip Cone Module for the Finnpipette* BioControl Electronic Pipettor, Variable Volume^ Volume: 50-300uL. Increments: 1.0uL. Accuracy: +-4.6 to 2.4%. Easy to remove. Interchangeable with single channel Biocontrol modules. Autoclavable.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BIOCONTROL TIP CONE 12CH 300UL 53515-962 2205690 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty