Would you like to log in again?
Accessories for Microtiter® 96-Well Plates, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Microtiter®
6604 9503130 5500 5701 5550 6305 3501
62402-929CS 295.6 USD InStock
62402-929 62403-072 62402-969 62402-936 62402-931 62402-921 53515-986
Accessories for Microtiter® 96-Well Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
These accessories are for use with all Thermo Scientific Immulon®, Microfluor®, and Microlite™ Microtiter plates and Removawell* assemblies.
Rigid styrene lids are sterile. Flexible vinyl lids 62403-072 are for use with Removawell assemblies. Clear precut acetate, Mylar®, and PVC plate sealers are for storage and incubation procedures.
The Removawell holder 62402-969 holds eight 12-well or twelve 8-well Removawell strips in a 96-well format.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty