Would you like to log in again?
Upchurch Scientific® Y Connectors, Multi-Port Connectors, IDEX Health & Science
Supplier: IDEX Health & Science
Upchurch Scientific®
P-513 P-514 P-512
53521-164EA 63.05 USD InStock
53521-164 53521-162 53521-160
Upchurch Scientific® Y Connectors, Multi-Port Connectors, IDEX Health & Science
Fittings and Connectors
Upchurch Scientific® PEEK™ Y Connectors are designed to split a stream or join two streams together, just like a tee
However, the configuration of a tee can lead to turbulent flow and solvent outgassing, which increases baseline noise and reduces sensitivity. The geometry of a Y connector creates much less turbulence and thus can improve analytical results. These standard Y Connectors use 1/4-28 Flangeless fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty