Would you like to log in again?
Mµlti™ PCR Tubes, Sorenson BioScience
Supplier: Sorenson BioScience
Mµlti™
16970 16990 16950 2319 16980
53550-114CS 1176.01 USD InStock
53550-114 53550-106 14229-856 53550-112 53550-110
Mµlti™ PCR Tubes, Sorenson BioScience
Tubes PCR Tubes
These 0.2 ml tubes fit most popular thermal cyclers.
Single polypropylene tubes with attached caps eliminate carry-over contamination and minimize evaporation. Thin-wall design ensures uniform rapid thermal transfer to reduce cycle times. Certified free of RNase, DNase, human DNA, and PCR inhibitors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty