Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MµltiFlex™ Pipette Tips, Sorenson BioScience

Supplier: Sorenson BioScience

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


MµltiFlex™
17310 13770 28480 13790 13760 13810
53550-190CS 293.95 USD InStock
53550-190 14229-864 53550-420 53550-167 53550-189 53550-178
MµltiFlex™ Pipette Tips, Sorenson BioScience
Pipette Tips
Ideal for loading gels and other discrete sampling applications.

  • A 5 µl graduation provides easy volume QuikCheck

Choose rounded-end tips with a 0.57 mm Ext.Ø for SDS PAGE gels, flat-ended tips with a 0.37 mm Ext.Ø for 0.4 mm sequencing gels, or flat-ended tips with a 0.17 mm Ext.Ø for 0.2 mm sequencing gels.

The 8.3 cm (3¹/₄") tip length permits easy access to narrow tubes and sample plates.

Tips provide positive displacement accuracy using standard air displacement pipettors. They fit most popular research-grade pipettors and feature a 33 mm flexible microcapillary that allows precision pipetting in the critical 0.5 to 200 µl range.

Certifications: Certified RNase- and DNase-free.

Ordering information: All tip orders are supplied in polypropylene racks of 200 tips, with the exception of item 14229-864, which is supplied as a bulk bag of 1000 tips.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty