Would you like to log in again?
Mµlti™ MiniFlex™ Pipette Tips, Sorenson BioScience
Supplier: Sorenson BioScience
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Mµlti™ MiniFlex™
15110 17350 15100 15130 17360 15120
53550-434CS 326.77 USD InStock
53550-434 53550-432 53550-428 53550-438 53550-436 53550-430
Mµlti™ MiniFlex™ Pipette Tips, Sorenson BioScience
Pipette Tips
Tips feature an elongated, flexible, ultra-thin microcapillary that is ideal for nucleic acid sequencing and protein separation.
  • A 2 µl graduation permits easy volume QuikCheck
  • Round-ended tip is ideal for SDS-PAGE gels
Flat-ended tip is specially designed for use in 0.4 and 0.2 mm sequencing gels. Tips have a unique universal sealing ring for use with most popular brand research-grade pipettors that deliver volumes from 0.1 to 10 µl.
Certifications: Certified RNase- and DNase-free.
Ordering information: Supplied in polypropylene MµltiRacks of 200 tips.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty