Would you like to log in again?
Microcentrifuge Tubes, Sorenson BioScience
Supplier: Sorenson BioScience
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
39640T 11500-E 11220 12030 11160
89082-332CS 227.63 USD InStock
89082-332 53550-937 53550-908 53550-792 53550-906
Microcentrifuge Tubes, Sorenson BioScience
Tubes Microcentrifuge Tubes
SafeSeal™ Microcentrifuge tubes are ideal for high-speed centrifugation, rated up to 25000×g.
  • Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors
A positive seal design allows repeated cap closures and will not pop during boiling or freezing. Tubes feature external graduation marks and frosted writing surfaces; fit all standard rotors; are autoclavable; and are rated from −80 to 121 °C.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty