Would you like to log in again?
MµlTI™ SafeSeal™ Microcentrifuge Tubes, Sorenson BioScience
Supplier: Sorenson BioScience
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MµlTI™ SafeSeal™
11210-E 16070 16590 16090 11300 16600 16140 16610 11700 16570 12160 16630 11590 16560 12000 12060 16550 16620
53550-930CS 376.71 USD InStock
53550-930 53550-698 53550-148 53550-960 53550-945 53550-965 53550-778 53550-920 53550-790 53550-970 53550-870 53550-916 53550-915 53550-925 53550-958 53550-935 53550-913 53550-917
MµlTI™ SafeSeal™ Microcentrifuge Tubes, Sorenson BioScience
Tubes Microcentrifuge Tubes
These disposable microtubes are designed for use in all popular microcentrifuges.
They are manufactured from precision molded polypropylene for biotechnology applications. The cap design allows repeated openings and closings yet resists popping from internal pressure or during boiling.
Delivery information: Tubes 53550-915 are supplied in individual bags of 200 of blue, yellow, violet, green, and red. Tubes 53550-778 are supplied in individual bags of 100 of blue, yellow, violet, green, and red. Tubes 53550-870 are supplied in individual bags of 80 of blue, yellow, violet, green, and red.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty