Would you like to log in again?
SAFETY SHIELD MODEL M
Supplier: CARVER INC.
Safety Shields^ Attractive cabinet encloses pressing area to protect operator from objects which might fly from the compression area. Hinged front access door is clear, shatterproof polycarbonate. Recommended for use on all presses. Will accommodate heater. For Model M press.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SAFETY SHIELD MODEL M 53880-076 3253 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty