Would you like to log in again?
SWIVEL PLATE F/HYDRAULIC PRESS
Supplier: CARVER INC.
Swivel Bearing Plate^ Requires safety shield. For Carver presses. Used for crushing and breaking tests of standard 5.1x5.1cm cubes and 5.1x10.2cm cylinders of concrete samples or similar crushing tests. Swivel Bearing Plates adjust slightly to the sample.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SWIVEL PLATE F/HYDRAULIC PRESS 53887-005 2092 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty