Would you like to log in again?
HAND PUMP POLYPROP TUBE
Supplier: RPR ZAHN , LLC.
Manual Pneumatic Hand Pump^ (MPP-H) Polypropylene tube and polypropylene pump body. Flows to 2gpm. Use for common acids and caustics.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HAND PUMP POLYPROP TUBE 54811-023 MPP-H Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty