Would you like to log in again?
DETERGENT AQUET PHOS-FRE 1GL
Supplier: Bel-Art Products, a Part of SP
Aquet* Phosphate-Free Liquid Detergent^ 3.8L, for removing film and grease from laboratory glassware and optics or as a wetting agent. Eliminates scrubbing and scraping. Diluted to a 1% solution, fast drying action eliminates wiping and drying. Biodegradable. Neutral pH. Non-ionic. Non-toxic.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DETERGENT AQUET PHOS-FRE 1GL 54848-002 F17094-0030 Case of 12 (1/ Itm/Bx) Retrieving
Each(1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty