Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Solvent-Based Paint Pen Markers, Bel-Art Products, a part of SP
Supplier: Bel-Art
Manomark™
F13377-0001 F13377-0000
54848-150PK 137.99 USD InStock
54848-150 54848-148
Solvent-Based Paint Pen Markers, Bel-Art Products, a part of SP
Pens Ink Pens
Writes on all plastics, glass, metal, wood, paper and cloth, including Teflon® fluoropolymer resins.
  • Precision seal valve provides smooth ink flow and preserves the pen’s life, even if left uncapped
  • When dry, the solvent based ink is temperature resistant from 0 to 155 °C (32 to 310 °F)
  • Markings are insoluble in water or hydrocarbons
Resists many solvents, but is easily removed with alcohol or acetone.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty