Would you like to log in again?
PUMP LIQ METERING 5000PSI 115V
Supplier: Eldex Corporation
Liquid Metering Pump^ Single-piston. Precise metering at high pressure is attained by a positive displacement, reciprocating piston assembly. 316 stainless steel construction. Flow rate: 0.05 to 1.5mL/min. Maximum pressure: 5000psi. Corrosion-resistant materials. 115V, 60Hz.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PUMP LIQ METERING 5000PSI 115V 54849-123 1003 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty